Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNRD1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551319

Catalog No. PA551319


Add to cart

Description

Description

The antibody detects endogenous levels of total ZNRD1 protein.

This gene encodes a DNA-directed RNA polymerase I subunit. The encoded protein contains two potential zinc-binding motifs and may play a role in regulation of cell proliferation. The encoded protein may be involved in cancer and human immunodeficiency virus progression. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

ZNRD1
Polyclonal
Unconjugated
ZNRD1
Liquid
ZNRD1
Full length fusion protein of human ZNRD1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
0.8 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q9P1U0
RPA12
Rabbit
IgG
100 μL
-20°C
Primary
30834
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit