Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNRF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | ZNRF1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB023372
|
Novus Biologicals
NBP294411100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB023373
|
Novus Biologicals
NBP29441120UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
ZNRF1 Polyclonal antibody specifically detects ZNRF1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
ZNRF1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
DKFZp434E229, EC 6.3.2, EC 6.3.2.-, FLJ14846, MGC15430, nerve injury gene 283, Nerve injury-induced gene 283 protein, nin283, NIN283E3 ubiquitin-protein ligase ZNRF1, zinc and ring finger 1, zinc and ring finger protein 1, Zinc/RING finger protein 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human ZNRF1 (NP_115644.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Zinc Finger | |
PBS (pH 7.3), 50% glycerol | |
84937 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title