Learn More
Alfa Aesar™ Zolmitriptan
A serotonin 5HT1D-receptor agonist
Manufacturer:  Alfa Aesar™ J60616MC
 View more versions of this product
Catalog No. AAJ60616MC
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
100mg
Crystalline Powder
139264-17-8
287.36g/mol
136° to 141°C
Signal Transduction Reagent
White
A serotonin 5HT1D-receptor agonist
C16H21N3O2
(S)-4-(3-[2-(Dimethylamino)ethyl]-5-indolylmethyl)oxazolidin-2-one
Soluble in DMSO at 5mg/ml