Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zolmitriptan

A serotonin 5HT1D-receptor agonist

$104.30 - $313.00

Specifications

CAS 139264-17-8
Formula Weight 287.36g/mol
Synonym (S)-4-(3-[2-(Dimethylamino)ethyl]-5-indolylmethyl)oxazolidin-2-one
Format Crystalline Powder
For Use With (Application) A serotonin 5HT1D-receptor agonist
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ60616MC View Documents Alfa Aesar™
J60616MC
100mg Each for $104.30
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ6061603 View Documents Alfa Aesar™
J6061603
1g Each for $313.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

139264-17-8
(S)-4-(3-[2-(Dimethylamino)ethyl]-5-indolylmethyl)oxazolidin-2-one
A serotonin 5HT1D-receptor agonist
136° to 141°C
Soluble in DMSO at 5mg/ml
287.36g/mol
Crystalline Powder
White
Signal Transduction Reagent
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit