Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zonisamide, USP, 98-101%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
68291-97-4
C8H8N2O3S
212.22
UBQNRHZMVUUOMG-UHFFFAOYSA-N
(1,2-benzoxazol-3-yl)methanesulfonamide
NS(=O)(=O)CC1=NOC2=CC=CC=C12
68291-97-4
C8H8N2O3S
212.22
UBQNRHZMVUUOMG-UHFFFAOYSA-N
(1,2-benzoxazol-3-yl)methanesulfonamide
NS(=O)(=O)CC1=NOC2=CC=CC=C12

Z1380, 68291-97-4, C8H8N2O3S

Spectrum™ Zonisamide, USP
Packaging Amber Glass Bottle
Ignition Residue 0.002
Quantity 100 g
Water 0.01
CAS Max % 1
Percent Purity 98 to 101%
Grade USP