Learn More
Swift SM105T-C-USB Zoom Stereo Microscope Digital Camera Package
Features microscope with variable zoom magnification range of 10X to 30X and Moticam 2.0 MP digital camera.
Manufacturer: Swift SM105TCUSB
Description
Microscope features:
- Energy-efficient variable, cordless, rechargeable illumination
- One-touch spring loaded stage clips
- Right eyepiece diopter focusing adjustment
Camera features:
- Model: D-Moticam 2
- Digital Interface: USB
- Live, digital output
- Detachable USB cable
- Feature-packed Motic Images Plus software
Specifications
Zoom Stereo Microscope Digital Camera Package | |
SM105T-C | |
W 10X, 20mm | |
6 Turret variable (0.06w) LED (Top variable: 0.05w, Bottom: 0.05w LED Cordless) |
Camera: 2.0 MP (1600 X 1200) | |
1X to 3X | |
Trinocular |