Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530879
Description
Recommended positive controls: NT2D1, IMR32, U87-MG. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The zona pellucida is an extracellular matrix that surrounds the oocyte and early embryo. It is composed primarily of three or four glycoproteins with various functions during fertilization and preimplantation development. The protein encoded by this gene is a structural component of the zona pellucida and functions in secondary binding and penetration of acrosome-reacted spermatozoa. The nascent protein contains a N-terminal signal peptide sequence, a conserved ZP domain, a consensus furin cleavage site, and a C-terminal transmembrane domain. It is hypothesized that furin cleavage results in release of the mature protein from the plasma membrane for subsequent incorporation into the zona pellucida matrix. However, the requirement for furin cleavage in this process remains controversial based on mouse studies.
Specifications
ZP2 | |
Polyclonal | |
Unconjugated | |
Zp2 | |
75-kDa rabbit zona pellucida protein; Processed zona pellucida sperm-binding protein 2; zona pellucida 2 glycoprotein; zona pellucida A protein; zona pellucida glycoprotein 2; zona pellucida glycoprotein 2 (sperm receptor); zona pellucida glycoprotein ZP2; zona pellucida glycoprotein ZPA; zona pellucida protein A; Zona pellucida sperm-binding protein 2; ZP2; Zp-2; ZPA | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7783 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.89 mg/mL | |
PBS with no preservative | |
Q05996 | |
Zp2 | |
Recombinant fragment corresponding to a region within amino acids 1 and 313 of Human ZP2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction