Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562907
Description
Immunogen sequence: QPLWLLQGGA SHPETSVQPV LVECQEATLM VMVSKDLFGT GKLIRAADLT LGPEACEPLV SMDTEDVVRF EVGLHECGNS MQVTD Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 58%.
The zona pellucida is an extracellular matrix that surrounds the oocyte and early embryo. It is composed primarily of three or four glycoproteins with various functions during fertilization and preimplantation development. The protein encoded by this gene is a structural component of the zona pellucida and functions in primary binding and induction of the sperm acrosome reaction. The nascent protein contains a N-terminal signal peptide sequence, a conserved ZP domain, a C-terminal consensus furin cleavage site, and a transmembrane domain. It is hypothesized that furin cleavage results in release of the mature protein from the plasma membrane for subsequent incorporation into the zona pellucida matrix. However, the requirement for furin cleavage in this process remains controversial based on mouse studies. A variation in the last exon of this gene has previously served as the basis for an additional ZP3 locus; however, sequence and literature review reveals that there is only one full-length ZP3 locus in the human genome. Another locus encoding a bipartite transcript designated POMZP3 contains a duplication of the last four exons of ZP3, including the above described variation, and maps closely to this gene.
Specifications
ZP3 | |
Polyclonal | |
Unconjugated | |
ZP3 | |
Processed zona pellucida sperm-binding protein 3; Sperm receptor; zona pellucida C; zona pellucida C protein; zona pellucida glycoprotein 3; zona pellucida glycoprotein 3 (sperm receptor); zona pellucida glycoprotein 3 preproprotein; zona pellucida glycoprotein 3A (sperm receptor); zona pellucida glycoprotein 3B; zona pellucida glycoprotein ZP3; zona pellucida glycoprotein ZP3B; zona pellucida protein C; Zona pellucida sperm-binding protein 3; Zp3; Zp-3; ZP3A; ZP3A/ZP3B; ZP3B; Zp-3B; ZPC | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7784 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P21754 | |
ZP3 | |
Recombinant protein corresponding to Human ZP3. Recombinant protein control fragment (Product #RP-101584). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction