Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZPBP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157768
Description
ZPBP2 Polyclonal specifically detects ZPBP2 in Human samples. It is validated for Western Blot.Specifications
ZPBP2 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
CAAX prenyl protease 1 homolog, EC 3.4.24.84, FACE1FLJ14968, FACE-1zinc metalloproteinase (STE24 homolog, yeast), Farnesylated proteins-converting enzyme 1, farnesylated-proteins converting enzyme 1, HGPS, Hutchinson-Gilford progeria syndrome, MADB, Prenyl protein-specific endoprotease 1, PRO1, Ste24p, STE24zinc metallopeptidase (STE24 homolog, yeast), zinc metallopeptidase (STE24 homolog, S. cerevisiae), Zinc metalloproteinase Ste24 homolog | |
Rabbit | |
Affinity Purified | |
RUO | |
124626 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
ZPBP2 | |
Synthetic peptides corresponding to ZPBP2 (zona pellucida binding protein 2) The peptide sequence was selected from the middle region of ZPBP2)(50ug). Peptide sequence VRLDSCRPGFGKNERLHSNCASCCVVCSPATFSPDVNVTCQTCVSVLTYG. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Human: 100%; Guinea pig: 92%; Equine: 92%; Mouse: 92%; Pig: 92%; Rat: 92%; Canine: 84%. | |
Human, Mouse, Rat, Equine, Guinea Pig, Rabbit | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title