Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZPBP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ZPBP2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP157768
|
Novus Biologicals
NBP157768 |
100 μL |
Each for $436.00
|
|
NBP15776820
|
Novus Biologicals
NBP15776820UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
ZPBP2 Polyclonal specifically detects ZPBP2 in Human samples. It is validated for Western Blot.Specifications
ZPBP2 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
124626 | |
Synthetic peptides corresponding to ZPBP2 (zona pellucida binding protein 2) The peptide sequence was selected from the middle region of ZPBP2)(50ug). Peptide sequence VRLDSCRPGFGKNERLHSNCASCCVVCSPATFSPDVNVTCQTCVSVLTYG. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
CAAX prenyl protease 1 homolog, EC 3.4.24.84, FACE1FLJ14968, FACE-1zinc metalloproteinase (STE24 homolog, yeast), Farnesylated proteins-converting enzyme 1, farnesylated-proteins converting enzyme 1, HGPS, Hutchinson-Gilford progeria syndrome, MADB, Prenyl protein-specific endoprotease 1, PRO1, Ste24p, STE24zinc metallopeptidase (STE24 homolog, yeast), zinc metallopeptidase (STE24 homolog, S. cerevisiae), Zinc metalloproteinase Ste24 homolog | |
ZPBP2 | |
IgG | |
Affinity Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title