Learn More
Aviva Systems Biology Corp ZSCAN12 Antibody - N-terminal region (ARP47528_T100) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47528_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZSCAN12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZSCAN12. Alternate names: KIAA0426, ZFP96, ZNF29K1, ZNF305, ZNF96, dJ29K1.2ApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-014-59578