Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZSCAN18 Mouse anti-Human, Clone: OTI4D10, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA505370

Catalog No. 50-167-5856


Add to cart

Description

Description

ZNF447 contains 1 SCAN box domain and 2 C2H2-type zinc fingers. It belongs to the kruppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.
Specifications

Specifications

ZSCAN18
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8TBC5
ZNF447
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
65982
Immunohistochemistry (Paraffin), Western Blot
OTI4D10
Unconjugated
ZSCAN18
Liquid
ZSCAN18
Full length human recombit protein of human ZSCAN18 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit