Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZSCAN2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548507
Description
May be involved in transcriptional regulation during the post-meiotic stages of spermatogenesis.Specifications
ZSCAN2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q7Z7L9 | |
ZSCAN2 | |
KLH conjugated synthetic peptide between 117-146 amino acids from the N-terminal region of human ZSCAN2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
ZSCAN2 | |
Zinc finger and SCAN domain-containing protein 2, Zinc finger protein 29 homolog, Zfp-29, Zinc finger protein 854, ZSCAN2, ZFP29, ZNF854 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54993 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction