Learn More
ZSCAN20 Rabbit anti-Human, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 208551AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-3122
Add to Cart
Description
Description
May be involved in transcriptional regulation.
Specifications
Specifications
ZSCAN20
Polyclonal
Unconjugated
ZSCAN20
KOX29, ZFP 31, Zinc finger protein 31, Zinc finger protein 360, Zinc finger protein KOX29, ZNF31, ZNF360, ZSCAN20
Rabbit
Antigen Affinity Chromatography
RUO
7579
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P17040
ZSCAN20
ZSCAN20 Fusion Protein Ag14798
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions