Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
ZSCAN21 Mouse anti-Canine, Human, Mouse, Clone: OTI2H8, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA506092
Description
ZSCAN21 belongs to the krueppel C2H2-type zinc-finger protein family and is a strong transcriptional activator.Specifications
ZSCAN21 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q07231, Q9Y5A6 | |
NY-REN-21, ZNF38, Zipro1 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
22697, 489848, 7589 |
Western Blot | |
OTI2H8 | |
Unconjugated | |
ZSCAN21 | |
Liquid | |
ZKSCAN1, Zscan21 | |
Full length human recombit protein of human ZSCAN21 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Canine, Human, Mouse |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title