Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZSCAN23 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549278

Catalog No. PA549278

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZSCAN23
Polyclonal
PBS with 0.09% sodium azide
Q3MJ62
Zinc finger and SCAN domain-containing protein 23, Zinc finger protein 390, Zinc finger protein 453, ZSCAN23, ZNF390, ZNF453
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
222696
Western Blot
Unconjugated
ZSCAN23
Liquid
ZSCAN23
KLH conjugated synthetic peptide between 200-229 amino acids from the Central region of human ZSCAN23.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit