Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZSCAN5 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310384100UL
Description
ZSCAN5 Polyclonal specifically detects ZSCAN5 in Human samples. It is validated for Western Blot.Specifications
ZSCAN5 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
MGC4161, zinc finger and SCAN domain containing 5, zinc finger and SCAN domain containing 5A, zinc finger and SCAN domain-containing protein 5A, Zinc finger protein 495ZSCAN5, ZNF495 | |
The immunogen is a synthetic peptide directed towards the middle region of Human ZSCAN5 (NP_077279). Peptide sequence QDSDIEMAEAPSSVRDDLKDVSSQRASSVNQMRPGEGQAHRELQILPRVP | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
79149 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction