Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZSCAN5B Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19141120UL

 View more versions of this product

Catalog No. NBP191420UL


Add to cart

Description

Description

ZSCAN5B Polyclonal antibody specifically detects Antigen in Human, Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

ZSCAN5B
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
zinc finger and SCAN domain containing 5B, zinc finger and SCAN domain-containing protein 5B, ZNF495B
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
342933
Western Blot
Western Blot 1:1000
Affinity Purified
ZSCAN5B
Synthetic peptide directed towards the C terminal of human hCG_2042202. Peptide sequence HKRIHTGERPFKCKYCSKVFSHKGNLNVHQRTHSGEKPYKCPTCQKAFRQ.
50 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit