Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZSWIM4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559861
Description
Immunogen sequence: PLDPIGCLCR ALLEACRLEE ETLTLYPDSG PEKRKVAYQH VPVPGSPGES YLVL Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 87%.

Specifications
ZSWIM4 | |
Polyclonal | |
Unconjugated | |
ZSWIM4 | |
Zinc finger SWIM domain-containing protein 4; zinc finger SWIM-type containing 4; zinc finger, SWIM domain containing 4; zinc finger, SWIM-type containing 4; ZSWIM4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
65249 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H7M6 | |
ZSWIM4 | |
Recombinant protein corresponding to Human ZSWIM4. Recombinant protein control fragment (Product #RP-99096). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction