Learn More
ZW10 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 245611AP
Catalog No. 50-173-4423
Add to cart
Description
Description
This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
Specifications
Specifications
ZW10
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43264
HZW10, KNTC1AP, ZW10
Rabbit
IgG
150 μL
-20°C
Primary
9183
Immunoprecipitation, Western Blot
Unconjugated
ZW10
Liquid
ZW10
ZW10 Fusion Protein Ag19138
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human