Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZWINT Mouse anti-Human, DyLight 488, Clone: 276, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243562G

Catalog No. NBP243562G


Add to cart

Description

Description

ZWINT Monoclonal antibody specifically detects ZWINT in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZWINT
276
Western Blot
Purified
ZWINT
Recombinant protein encompassing a sequence within the center region of human ZWINT. The exact sequence is proprietary.
Protein G purified
Cell Cycle and Replication, DNA Repair, Mitotic Regulators
Primary
11130
Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
C20orf164, chromosome 20 open reading frame 164, dJ337O18.7, zinc finger SWIM domain-containing protein 3, zinc finger, SWIM domain containing 3, zinc finger, SWIM-type containing 3
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit