Learn More
Sklar InstrumentsSupplier Diversity Partner Obwegeser Zygomatic Arch Awl for Wire Suturing, Sklar, 9 in., Sharp Fenestrated, Slightly Curved, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 262009
Obwegeser Zygomatic Arch Awl for Wire Suturing, Sklar, 9 in., Sharp Fenestrated, Slightly Curved, Stainless Steel. Sklar's Obwegeser Zygomatic Arch Awl is used for wire suturing in oral procedures involving the cheekbone.
Catalog No. 50-143-8084