Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Zymosan, Saccharomyces cerevisiae

Click to view available options
Quantity:
250mg
1g
58856-93-2
MFCD00082157
58856-93-2
MFCD00082157
An insoluble preparation of yeast cell, has been shown to activate macrophages via toll-like receptor 2.
Color White
Quantity 1g
Merck Index 14,10200
Physical Form Powder
Chemical Name or Material Zymosan, Saccharomyces cerevisiae

RTECSNumber : ZI2750000

Recommended Storage : 2° to 8°C