Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZZZ3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565811
Description
Immunogen sequence: QMQAESGFVQH VGFKCDNCGI EPIQGVRWHC QDCPPEMSLD FCDSCSDCLH ETDIHKEDHQ LEPIYRSETF LDRDYCVSQG TSYNYLDPNY FPAN Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.
Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.
Specifications
ZZZ3 | |
Polyclonal | |
Unconjugated | |
ZZZ3 | |
3110065C23Rik; 6430567E01Rik; AA408566; AI481057; ATAC component 1 homolog; ATAC1; AW556059; DKFZP564I052; R75514; RGD1565468; zinc finger ZZ-type containing 3; zinc finger, ZZ domain containing 3; zinc finger, ZZ-type containing 3; ZZ-type zinc finger-containing protein 3; Zzz3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26009 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IYH5 | |
ZZZ3 | |
Recombinant Human ZZZ3. Recombinant protein control fragment (Product #RP-106485). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction