Fisherbrand™ Class B Amber Glass Threaded Vials with Attached Caps
| Description | Catalog Number | Available Packaging |
|---|---|---|
| Capacity: 4mL; 15 O.D x 45mm H |
14-955-331
NO.:90J14-955-331
|
Case of 144 Each |
| Capacity: 2mL; 12 O.D x 35mm H |
14-955-321
NO.:90G14-955-321
|
Case of 288 Each |
| Capacity: 40mL; 28 O.D x 95mm H |
14-955-322
NO.:90X14-955-322
|
Case of 72 Each |
| Capacity: 8mL; 17 O.D x 60mm H |
14-955-332
NO.:90L14-955-332
|
Case of 144 Each |
| Capacity: 20mL; 28 O.D x 57mm H |
14-955-333
NO.:90X14-955-333
|
Case of 72 Each |
Key Features:
- Threaded
- Attached black phenolic screw-top closures with PTFE-faced white 14B rubber liners
Compliance:
Meet ASTM™ Type I, Class B and USP Type I standardsSpecifications:
| Classification | B |
|---|---|
| Material (Liner) | Teflon-faced Rubber |
| Color | Amber |
| Material | Amber Glass (Class B) |
| For Use With (Application) | For general purpose vials for light sensitive materials |
| Closure Material | Phenolic |
| Closure Type | Fitted Cap |
| Autoclavable | Autoclavable |
| Certifications/Compliance | ASTM, USP Type I |
| Product Type | Vial |
//assets.adobedtm.com/7835b01639d4/846649467620/launch-40ac9df91374.min.js
3972700
hosted-catalog:product:-Fisherbrandtradenbsp-Class-B-Amber-Glass-Threaded-Vialswith-Attached-Caps|3972700
14955331