This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02157892_g1
See other HIK GE Assays ›
Gene Symbol:
HIK
Gene Name:
ATP binding microtubule motor family protein
Gene Aliases:
ARABIDOPSIS NPK1-ACTIVATING KINESIN 1, AT1G18370, ATNACK1, F15H18.12, F15H18_12, HINKEL, NACK1, NPK1-ACTIVATING KINESIN 1
Chromosome Location:
Chr.1: 6319450 - 6324378 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02157892_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HIK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_101695.5 NP_173273.2 12 - 13 1566 76
GenBank mRNA AB081599.1 - 11 - 12 1299 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cytokinesis by cell plate formation
microtubule-based movement
pollen development
embryo sac cellularization
gametophyte development
microtubule motor activity
microtubule binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-c7pcq:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0