This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02158766_gH
See other AT1G19660 GE Assays ›
Gene Symbol:
AT1G19660
Gene Name:
Wound-responsive family protein
Gene Aliases:
AT1G19660, AtBBD2, BBD2, F14P1.1, F14P1_1, bifunctional nuclease in basal defense response 2
Chromosome Location:
Chr.1: 6799969 - 6802906 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02158766_gH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT1G19660
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035991.1 NP_001031068.1 2 - 3 472 92
NM_001332402.1 NP_001322963.1 2 - 3 514 92
NM_101822.4 NP_564093.1 2 - 3 648 92
GenBank mRNA AK317508.1 - 2 - 3 473 92
AY035016.1 - 2 - 3 391 92
AY059082.1 - 1 - 2 296 92

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline