This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02158772_g1
See other AT1G19660 GE Assays ›
Gene Symbol:
AT1G19660
Gene Name:
Wound-responsive family protein
Gene Aliases:
AT1G19660, AtBBD2, BBD2, F14P1.1, F14P1_1, bifunctional nuclease in basal defense response 2
Chromosome Location:
Chr.1: 6799969 - 6802906 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02158772_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AT1G19660
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035991.1 NP_001031068.1 8 - 9 1010 107
NM_001332402.1 NP_001322963.1 8 - 9 1052 107
NM_101822.4 NP_564093.1 8 - 9 1186 107
GenBank mRNA AK317508.1 - 8 - 9 1011 107
AY035016.1 - 8 - 9 929 107
AY059082.1 - 7 - 8 834 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9mkk8:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0