This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02159323_g1
See other CAT3 GE Assays ›
Gene Symbol:
CAT3
Gene Name:
catalase 3
Gene Aliases:
AT1G20620, ATCAT3, F5M15.5, F5M15_5, SEN2, SENESCENCE 2, catalase 3
Chromosome Location:
Chr.1: 7142972 - 7146540 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02159323_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CAT3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001035995.4 NP_001031072.3 1 - 2 1170 96
NM_001035996.1 NP_001031073.1 2 - 3 186 96
NM_001332452.1 NP_001323332.1 2 - 3 1069 96
NM_101913.4 NP_564120.1 2 - 3 281 96
NM_202144.2 NP_973873.1 2 - 3 186 96
GenBank mRNA AK316990.1 - 2 - 3 191 96
AY056447.1 - 2 - 3 165 96
AY058104.1 - 2 - 3 168 96

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001035995.4 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
cellular response to nitrogen starvation
response to cold
response to light stimulus
cellular response to sulfate starvation
cellular response to phosphate starvation
peptidyl-cysteine S-trans-nitrosylation
response to hydrogen peroxide
hydrogen peroxide catabolic process
response to cadmium ion
oxidation-reduction process
mRNA binding
catalase activity
protein binding
heme binding
peptidyl-cysteine S-nitrosylase activity
cobalt ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0