This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02161345_g1
See other PIN4 GE Assays ›
Gene Symbol:
PIN4
Gene Name:
Auxin efflux carrier family protein
Gene Aliases:
ARABIDOPSIS PIN-FORMED 4, AT2G01420, ATPIN4, AUXIN TRANSPORTER SPLICE VARIANT B, F10A8.27, PIN-FORMED 4
Chromosome Location:
Chr.2: 180033 - 183764 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02161345_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PIN4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001335061.1 NP_001323732.1 4 - 5 2596 148
NM_126203.3 NP_565261.1 5 - 6 2332 148
NM_179592.2 NP_849923.1 5 - 6 2344 148
GenBank mRNA AF087016.1 - 5 - 6 1803 148
AY091009.1 - 5 - 6 1903 148
BT008691.1 - 5 - 6 1908 148

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

pattern specification process
auxin polar transport
auxin homeostasis
auxin efflux
root development
transmembrane transport
auxin efflux transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zl4nj:80/100.66.77.81:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0