This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02161917_g1
See other GSTZ1 GE Assays ›
Gene Symbol:
GSTZ1
Gene Name:
glutathione S-transferase zeta 1
Gene Aliases:
AT2G02390, ATGSTZ1, AtMAAI, GLUTATHIONE S-TRANSFERASE 18, GST18, MAAI, T16F16.18, T16F16_18, glutathione S-transferase zeta 1, maleylacetoacetate isomerase
Chromosome Location:
Chr.2: 628730 - 631165 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02161917_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTZ1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001335107.1 NP_001325193.1 1 - 2 261 96
NM_126296.5 NP_178344.1 4 - 5 522 96
NM_179595.5 NP_849926.2 4 - 5 353 96
NM_201671.3 NP_973400.1 4 - 5 396 96
GenBank mRNA AF288182.1 - 3 - 4 237 96
AJ278293.1 - 3 - 4 237 96
AK226342.1 - 4 - 5 307 96
AY052332.1 - 4 - 5 290 96
AY061901.1 - 3 - 4 237 96
AY208155.1 - 4 - 5 288 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

L-phenylalanine catabolic process
glutathione metabolic process
aromatic amino acid family metabolic process
toxin catabolic process
homogentisate catabolic process
glutathione transferase activity
maleylacetoacetate isomerase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0