Assay Details
| Gene Symbol: PEX19-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111220.4 | NP_186999.1 | 1 - 2 | 284 | 92 | |
| GenBank mRNA | AJ564199.1 | - | 1 - 2 | 151 | 92 | |
| BT012633.1 | - | 1 - 2 | 46 | 92 | ||
More Information
Set Membership: |
|

PEX19-1
Genomic Map