This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02164011_g1
See other CAT4 GE Assays ›
Gene Symbol:
CAT4
Gene Name:
cationic amino acid transporter 4
Gene Aliases:
AT3G03720, F20H23.25, F20H23_25, cationic amino acid transporter 4
Chromosome Location:
Chr.3: 925556 - 930002 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02164011_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CAT4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161114.2 NP_001154586.1 6 - 7 1049 109
GenBank mRNA AY062508.1 - 6 - 7 962 109
BT008369.1 - 6 - 7 747 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transport
amino acid transmembrane transporter activity
basic amino acid transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0