This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02167228_g1
See other ABCG43 GE Assays ›
Gene Symbol:
ABCG43 ABCG42
Gene Name:
ABC-2 and Plant PDR ABC-type transporter family protein
ABC-2 and Plant PDR ABC-type transporter family protein
Gene Aliases:
AT4G15236, ATP-binding cassette G43
AT4G15233, ATP-binding cassette G42
Chromosome Location:
Chr.4: 8696677 - 8702727 on Build TAIR10.1
Chromosome Location:
Chr.4: 8687354 - 8694733 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
-
Assay ID At02167228_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCG43
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_148328.3 NP_680694.3 7 - 8 873 93
Gene Symbol: ABCG42
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203808.2 NP_001190737.1 7 - 8 912 93
NM_001341013.1 NP_001319945.1 7 - 8 951 93
NM_001341014.1 NP_001329434.1 9 - 10 1407 93
NM_001341017.1 NP_001329431.1 9 - 10 1043 93
NM_001341019.1 NP_001329435.1 7 - 8 897 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Process(es)

ATPase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0