This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02169399_g1
See other ROPGEF14 GE Assays ›
Gene Symbol:
ROPGEF14
Gene Name:
RHO guanyl-nucleotide exchange factor 14
Gene Aliases:
AT1G31650, ATROPGEF14, F27M3.15, F27M3_15, RHO guanyl-nucleotide exchange factor 14, ROP (rho of plants) guanine nucleotide exchange factor 14
Chromosome Location:
Chr.1: 11326251 - 11330791 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02169399_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ROPGEF14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001332971.1 NP_001320846.1 3 - 4 960 98
NM_102900.5 NP_174446.2 5 - 6 1161 98
GenBank mRNA AK222010.1 - 5 - 6 1108 98

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of pollen tube growth
Rho guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9mkk8:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0