This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02169735_g1
See other BZIP68 GE Assays ›
Gene Symbol:
bZIP68
Gene Name:
basic region/leucine zipper transcription factor 68
Gene Aliases:
AT1G32150, AtbZIP68, F3C3.7, F3C3_7, basic region/leucine zipper transcription factor 68
Chromosome Location:
Chr.1: 11565494 - 11568645 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02169735_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: bZIP68
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_102948.4 NP_174494.2 12 - 13 1331 141
GenBank mRNA BT004147.1 - 12 - 13 1259 141

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
protein binding
identical protein binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0