Assay Details
| Gene Symbol: BOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084602.2 | NP_001078071.1 | 2 - 3 | 824 | 64 | |
| NM_180138.3 | NP_850469.1 | 3 - 4 | 915 | 64 | ||
| GenBank mRNA | AB073713.1 | - | 3 - 4 | 374 | 64 | |
| AY070067.1 | - | 3 - 4 | 730 | 64 | ||
| AY096436.1 | - | 3 - 4 | 374 | 64 | ||
| BT000732.1 | - | 3 - 4 | 730 | 64 | ||
More Information
Set Membership: |
|

BOR1
Genomic Map