Assay Details
| Gene Symbol: HDA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001338339.1 | NP_001326233.1 | 6 - 7 | 735 | 81 | |
| NM_001338340.1 | NP_001326232.1 | 6 - 7 | 735 | 81 | ||
| NM_112737.3 | NP_566612.1 | 6 - 7 | 752 | 81 | ||
| NM_180278.3 | NP_850609.2 | 6 - 7 | 735 | 81 | ||
More Information
Set Membership: |
|

HDA15
Genomic Map