Assay Details
| Gene Symbol: GLTP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161160.2 | NP_001154632.1 | 5 - 6 | 853 | 80 | |
| NM_113021.3 | NP_566679.1 | 3 - 4 | 831 | 80 | ||
| NM_180288.2 | NP_850619.1 | 4 - 5 | 673 | 80 | ||
| GenBank mRNA | AK228318.1 | - | 4 - 5 | 486 | 80 | |
More Information
Set Membership: |
|

GLTP3
Genomic Map