This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02174358_g1
See other PPA1 GE Assays ›
Gene Symbol:
PPa1
Gene Name:
pyrophosphorylase 1
Gene Aliases:
AT1G01050, AtPPa1, T25K16.5, T25K16_5, pyrophosphorylase 1
Chromosome Location:
Chr.1: 31170 - 33171 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02174358_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPa1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001331245.1 NP_001322481.1 8 - 9 764 66
NM_099987.4 NP_171613.1 8 - 9 736 66
GenBank mRNA AB493418.1 - 7 - 8 593 66
AK226833.1 - 8 - 9 683 66
AY052717.1 - 8 - 9 679 66
AY057668.1 - 8 - 9 682 66
AY065201.1 - 8 - 9 683 66
AY081555.1 - 7 - 8 593 66
AY085015.1 - 8 - 9 713 66
BT001144.1 - 7 - 8 593 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

pyrophosphatase

Gene Ontology Categories:

Function(s) Process(es)

sucrose metabolic process
phosphate-containing compound metabolic process
lipid storage
cell wall biogenesis
regulation of starch metabolic process
magnesium ion binding
inorganic diphosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline