Assay Details
| Gene Symbol: ROPGEF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120279.3 | NP_195821.1 | 3 - 4 | 816 | 92 | |
| GenBank mRNA | BT015831.1 | - | 3 - 4 | 638 | 92 | |
| BT020213.1 | - | 3 - 4 | 476 | 92 | ||
More Information
Set Membership: |
|

ROPGEF7
Genomic Map