This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02182859_g1
See other APC3 GE Assays ›
Gene Symbol:
APC3
Gene Name:
Mitochondrial substrate carrier family protein
Gene Aliases:
AT5G07320, ATP/phosphate carrier 3, T2I1.30, T2I1_30
Chromosome Location:
Chr.5: 2309863 - 2312346 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02182859_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_120814.3 NP_196349.1 1 - 2 783 69
GenBank mRNA AK175228.1 - 1 - 2 735 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

mitochondrial carrier protein

Gene Ontology Categories:

Function(s) Process(es)

drug transmembrane transport
ADP transport
ATP transport
phosphate ion transmembrane transport
calcium ion transmembrane transport
AMP transport
cation transmembrane transport
anion transmembrane transport
nucleotide transmembrane transport
ATP transmembrane transporter activity
calcium ion binding
calcium ion transmembrane transporter activity
phosphate ion transmembrane transporter activity
ADP transmembrane transporter activity
AMP transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0