Assay Details
| Gene Symbol: ACAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124198.4 | NP_568694.2 | 3 - 4 | 450 | 65 | |
| NM_180823.3 | NP_851154.1 | 2 - 3 | 520 | 65 | ||
| GenBank mRNA | AF364059.1 | - | 3 - 4 | 319 | 65 | |
| AK317237.1 | - | 3 - 4 | 223 | 65 | ||
| AY059736.1 | - | 1 - 2 | 188 | 65 | ||
| AY091271.1 | - | 1 - 2 | 117 | 65 | ||
More Information
Set Membership: |
|

ACAT2
Genomic Map