This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02186283_g1
See other ACAT2 GE Assays ›
Gene Symbol:
ACAT2
Gene Name:
acetoacetyl-CoA thiolase 2
Gene Aliases:
AACT2, AT5G48230, Acetoacetyl-CoA thiolase 2, EMB1276, EMBRYO DEFECTIVE 1276, MIF21.12, MIF21_12, acetoacetyl-CoA thiolase 2
Chromosome Location:
Chr.5: 19552142 - 19555531 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02186283_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_124198.4 NP_568694.2 5 - 6 729 112
NM_180823.3 NP_851154.1 4 - 5 799 112
GenBank mRNA AF364059.1 - 5 - 6 598 112
AK317237.1 - 5 - 6 502 112
AY059736.1 - 3 - 4 467 112
AY091271.1 - 3 - 4 396 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

fatty acid beta-oxidation
embryo development ending in seed dormancy
pollen germination
pollen tube growth
sterol metabolic process
acetyl-CoA C-acetyltransferase activity
acetyl-CoA C-acyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0