This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02186503_m1
See other CNGC4 GE Assays ›
Gene Symbol:
CNGC4
Gene Name:
cyclic nucleotide-gated cation channel 4
Gene Aliases:
AT5G54250, ATCNGC4, DEFENSE, DND2, HLM1, MDK4.7, MDK4_7, NO DEATH 2, cyclic nucleotide-gated cation channel 4
Chromosome Location:
Chr.5: 22025035 - 22030099 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02186503_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNGC4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203607.1 NP_001190536.1 1 - 2 731 114
NM_001345080.1 NP_001332114.1 1 - 2 731 114
NM_124805.3 NP_200236.1 1 - 2 692 114
NM_180857.2 NP_851188.1 1 - 2 712 114
GenBank mRNA AK316810.1 - 1 - 2 688 114
AY057691.1 - 1 - 2 661 114
Y17912.1 - 603 114

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ion channel ligand-gated ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

plant-type hypersensitive response
ion channel activity
calmodulin binding
cation transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0