This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02186504_m1
See other CNGC4 GE Assays ›
Gene Symbol:
CNGC4
Gene Name:
cyclic nucleotide-gated cation channel 4
Gene Aliases:
AT5G54250, ATCNGC4, DEFENSE, DND2, HLM1, MDK4.7, MDK4_7, NO DEATH 2, cyclic nucleotide-gated cation channel 4
Chromosome Location:
Chr.5: 22025035 - 22030099 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02186504_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNGC4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203607.1 NP_001190536.1 2 - 3 948 80
NM_001345080.1 NP_001332114.1 2 - 3 948 80
NM_124805.3 NP_200236.1 2 - 3 909 80
NM_180857.2 NP_851188.1 2 - 3 929 80
GenBank mRNA AK316810.1 - 2 - 3 905 80
AY057691.1 - 2 - 3 878 80
Y17912.1 - 820 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ion channel ligand-gated ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

plant-type hypersensitive response
ion channel activity
calmodulin binding
cation transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline