This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02186505_g1
See other CNGC4 GE Assays ›
Gene Symbol:
CNGC4
Gene Name:
cyclic nucleotide-gated cation channel 4
Gene Aliases:
AT5G54250, ATCNGC4, DEFENSE, DND2, HLM1, MDK4.7, MDK4_7, NO DEATH 2, cyclic nucleotide-gated cation channel 4
Chromosome Location:
Chr.5: 22025035 - 22030099 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02186505_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNGC4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203607.1 NP_001190536.1 3 - 4 1258 89
NM_001345080.1 NP_001332114.1 3 - 4 1258 89
NM_124805.3 NP_200236.1 3 - 4 1219 89
NM_180857.2 NP_851188.1 3 - 4 1239 89
GenBank mRNA AK316810.1 - 3 - 4 1215 89
AY057691.1 - 3 - 4 1188 89
Y17912.1 - 1130 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ion channel ligand-gated ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

plant-type hypersensitive response
ion channel activity
calmodulin binding
cation transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0