This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02186508_g1
See other CNGC4 GE Assays ›
Gene Symbol:
CNGC4
Gene Name:
cyclic nucleotide-gated cation channel 4
Gene Aliases:
AT5G54250, ATCNGC4, DEFENSE, DND2, HLM1, MDK4.7, MDK4_7, NO DEATH 2, cyclic nucleotide-gated cation channel 4
Chromosome Location:
Chr.5: 22025035 - 22030099 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02186508_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNGC4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001203607.1 NP_001190536.1 6 - 7 1675 102
NM_001345080.1 NP_001332114.1 6 - 7 1690 102
NM_124805.3 NP_200236.1 6 - 7 1651 102
NM_180857.2 NP_851188.1 6 - 7 1671 102
GenBank mRNA AK316810.1 - 6 - 7 1647 102
AY057691.1 - 6 - 7 1620 102
Y17912.1 - 1562 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons

Panther Classification:

Molecular Function -

ion channel ligand-gated ion channel transporter

Gene Ontology Categories:

Function(s) Process(es)

plant-type hypersensitive response
ion channel activity
calmodulin binding
cation transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0