This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › At02186839_g1
See other RPE GE Assays ›
Gene Symbol:
RPE
Gene Name:
D-ribulose-5-phosphate-3-epimerase
Gene Aliases:
AT5G61410, D-RIBULOSE-5-PHOSPHATE-3-EPIMERASE, D-ribulose-5-phosphate-3-epimerase, EMB2728, EMBRYO DEFECTIVE 2728
Chromosome Location:
Chr.5: 24683606 - 24686217 on Build TAIR10.1
Species:
Arabidopsis
Species Specific ID (Flybase ID):
-
Assay ID At02186839_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_125534.4 NP_200949.1 2 - 3 501 67
NM_180909.3 NP_851240.1 2 - 3 609 67
GenBank mRNA AF015274.1 - 2 - 3 333 67
AY045855.1 - 2 - 3 360 67
AY091381.1 - 2 - 3 228 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

pentose-phosphate shunt, non-oxidative branch
response to cold
response to nematode
embryo development ending in seed dormancy
reductive pentose-phosphate cycle
pentose catabolic process
cellular carbohydrate metabolic process
ribulose-phosphate 3-epimerase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline