Assay Details
| Gene Symbol: TSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115321.4 | NP_567004.1 | 7 - 8 | 1027 | 63 | |
| GenBank mRNA | BT025875.1 | - | 7 - 8 | 807 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSA1| Gene Symbol: TSA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115321.4 | NP_567004.1 | 7 - 8 | 1027 | 63 | |
| GenBank mRNA | BT025875.1 | - | 7 - 8 | 807 | 63 | |
Set Membership: |
|